Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LIX1L
Entrez Gene ID
106501930
Gene Name
limb and CNS expressed 1 like
Gene Aliases
-
Chromosome Location
Chr.3: 99092973 - 99113911 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_013962464.2 XP_013817918.2 5 - 6 877 65

View Full Product Details