Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MSS51
Entrez Gene ID
102184500
Gene Name
MSS51 mitochondrial translational activator
Gene Aliases
-
Chromosome Location
Chr.28: 16207069 - 16217097 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005699206.3 XP_005699263.2 5 - 6 3201 77

View Full Product Details