Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NAMPT
Entrez Gene ID
102173810
Gene Name
nicotinamide phosphoribosyltransferase
Gene Aliases
-
Chromosome Location
Chr.4: 72928491 - 72971901 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018047232.1 XP_017902721.1 4 - 5 787 65
GenBank mRNA JT656885.1 - 7 - 8 1341 65

View Full Product Details