Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KCND3
Entrez Gene ID
102175822
Gene Name
potassium voltage-gated channel subfamily D member 3
Gene Aliases
-
Chromosome Location
Chr.3: 89068171 - 89293460 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018045915.1 XP_017901404.1 7 - 8 2026 60
XM_018045916.1 XP_017901405.1 7 - 8 2101 60
XM_018045917.1 XP_017901406.1 6 - 7 1969 60
GenBank mRNA JT669658.1 - 9 - 10 2461 60

View Full Product Details