Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MYBPHL
Entrez Gene ID
102170477
Gene Name
myosin binding protein H like
Gene Aliases
-
Chromosome Location
Chr.3: 86415219 - 86429978 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005677958.2 XP_005678015.2 4 - 5 677 53

View Full Product Details