Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PTHLH
Entrez Gene ID
100860907
Gene Name
parathyroid hormone like hormone
Gene Aliases
PTHRP
Chromosome Location
Chr.5: 80443977 - 80458623 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001285753.1 NP_001272682.1 1 - 2 99 95
XM_013964082.2 XP_013819536.1 2 - 3 445 95
XM_013964083.2 XP_013819537.1 3 - 4 577 95
XM_018047539.1 XP_017903028.1 2 - 3 523 95
XM_018047541.1 XP_017903030.1 3 - 4 548 95
XM_018047542.1 XP_017903031.1 3 - 4 532 95
XM_018047543.1 XP_017903032.1 2 - 3 508 95
GenBank mRNA GU573787.1 - 1 - 2 99 95
KA309734.1 - 3 - 4 414 95

View Full Product Details