Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SPACA3
Entrez Gene ID
100860900
Gene Name
sperm acrosome associated 3
Gene Aliases
SLLP1
Chromosome Location
Chr.19: 16907196 - 16917523 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001286975.1 NP_001273904.1 1 - 2 230 64
XM_018063567.1 XP_017919056.1 2 - 3 406 64
GenBank mRNA HM038035.1 - 1 - 2 230 64

View Full Product Details