Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SPA17
Entrez Gene ID
102181367
Gene Name
sperm autoantigenic protein 17
Gene Aliases
-
Chromosome Location
Chr.29: 27486578 - 27499001 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005699579.3 XP_005699636.1 3 - 4 459 88

View Full Product Details