Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
EDN3
Entrez Gene ID
102174855
Gene Name
endothelin 3
Gene Aliases
-
Chromosome Location
Chr.13: 56388627 - 56413179 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018057604.1 XP_017913093.1 2 - 3 724 71

View Full Product Details