Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SNX16
Entrez Gene ID
102174761
Gene Name
sorting nexin 16
Gene Aliases
-
Chromosome Location
Chr.14: 1427744 - 1461757 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018058161.1 XP_017913650.1 7 - 8 1146 81
XM_018058162.1 XP_017913651.1 7 - 8 1125 81
XM_018058163.1 XP_017913652.1 7 - 8 1096 81
XM_018058164.1 XP_017913653.1 7 - 8 1063 81
XM_018058165.1 XP_017913654.1 8 - 9 1059 81
GenBank mRNA JT697543.1 - 7 - 8 1010 81

View Full Product Details