Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GALNT1
Entrez Gene ID
102174078
Gene Name
polypeptide N-acetylgalactosaminyltransferase 1
Gene Aliases
-
Chromosome Location
Chr.24: 21684611 - 21763071 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005696978.3 XP_005697035.1 11 - 12 1711 67

View Full Product Details