Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZNRF2
Entrez Gene ID
102170112
Gene Name
zinc and ring finger 2
Gene Aliases
-
Chromosome Location
Chr.4: 54186583 - 54278605 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018047106.1 XP_017902595.1 2 - 3 1611 114
GenBank mRNA JO437490.2 - 2 - 3 443 114

View Full Product Details