Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SACS
Entrez Gene ID
102169306
Gene Name
sacsin molecular chaperone
Gene Aliases
sacsin
Chromosome Location
Chr.12: 52089916 - 52171305 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018056556.1 XP_017912045.1 9 - 10 2802 92

View Full Product Details