Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AMACR
Entrez Gene ID
102185397
Gene Name
alpha-methylacyl-CoA racemase
Gene Aliases
-
Chromosome Location
Chr.20: 39726942 - 39742409 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018065752.1 XP_017921241.1 3 - 4 648 94

View Full Product Details