Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KLHL33
Entrez Gene ID
102179008
Gene Name
kelch like family member 33
Gene Aliases
-
Chromosome Location
Chr.10: 76288947 - 76298491 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005685411.3 XP_005685468.2 1 - 2 95 58

View Full Product Details