Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HOXA6
Entrez Gene ID
102170296
Gene Name
homeobox A6
Gene Aliases
-
Chromosome Location
Chr.4: 51314373 - 51319004 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005679275.3 XP_005679332.1 1 - 2 2853 56
GenBank mRNA JO517952.2 - - 217 56

View Full Product Details