Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CDK2AP1
Entrez Gene ID
102184835
Gene Name
cyclin dependent kinase 2 associated protein 1
Gene Aliases
-
Chromosome Location
Chr.17: 18896721 - 18907299 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018061356.1 XP_017916845.1 2 - 3 477 96
GenBank mRNA JO438004.2 - 2 - 3 902 96
JO502210.2 - 2 - 3 404 96

View Full Product Details