Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
SURF6
Entrez Gene ID
102173754
Gene Name
surfeit 6
Gene Aliases
-
Chromosome Location
Chr.11: 103647730 - 103654911 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018056139.1 XP_017911628.1 1 - 2 270 57
GenBank mRNA JO431066.2 - 1 - 2 221 57

View Full Product Details