Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CPEB2
Entrez Gene ID
102169571
Gene Name
cytoplasmic polyadenylation element binding protein 2
Gene Aliases
-
Chromosome Location
Chr.6: 110429923 - 110508618 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018049737.1 XP_017905226.1 4 - 5 2136 103
XM_018049738.1 XP_017905227.1 4 - 5 2136 103
XM_018049740.1 XP_017905229.1 3 - 4 2046 103
XM_018049741.1 XP_017905230.1 3 - 4 2046 103
GenBank mRNA JO325351.2 - 2 - 3 119 103

View Full Product Details