Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GRK7
Entrez Gene ID
102180541
Gene Name
G protein-coupled receptor kinase 7
Gene Aliases
-
Chromosome Location
Chr.1: 126668121 - 126709853 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005675490.2 XP_005675547.2 2 - 3 1103 103

View Full Product Details