Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLFN14
Entrez Gene ID
102184655
Gene Name
schlafen family member 14
Gene Aliases
-
Chromosome Location
Chr.19: 14422825 - 14435521 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005693334.2 XP_005693391.2 1 - 2 1073 115

View Full Product Details