Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CDCP2
Entrez Gene ID
102188732
Gene Name
CUB domain containing protein 2
Gene Aliases
-
Chromosome Location
Chr.3: 28695901 - 28707805 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018046499.1 XP_017901988.1 3 - 4 1093 107

View Full Product Details