Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
OXGR1
Entrez Gene ID
102189139
Gene Name
oxoglutarate receptor 1
Gene Aliases
-
Chromosome Location
Chr.12: 12124637 - 12125650 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005687762.2 XP_005687819.2 1 - 1 920 88

View Full Product Details