Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PIGT
Entrez Gene ID
102175138
Gene Name
phosphatidylinositol glycan anchor biosynthesis class T
Gene Aliases
-
Chromosome Location
Chr.13: 73224687 - 73236148 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005688593.3 XP_005688650.2 7 - 8 952 63

View Full Product Details