Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MID2
Entrez Gene ID
102174818
Gene Name
midline 2
Gene Aliases
-
Chromosome Location
Chr.X|NW_017189517: 35621935 - 35648810 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018044261.1 XP_017899750.1 - 425 64
GenBank mRNA JO445265.2 - - 425 64

View Full Product Details