Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRIM38
Entrez Gene ID
100860991
Gene Name
tripartite motif containing 38
Gene Aliases
-
Chromosome Location
Chr.23: 18042527 - 18057460 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005696760.3 XP_005696817.1 2 - 3 560 68
XM_005696762.3 XP_005696819.1 3 - 4 606 68
XM_018039208.1 XP_017894697.1 2 - 3 670 68
XM_018039209.1 XP_017894698.1 2 - 3 665 68
GenBank mRNA JI861635.1 - 1 - 2 401 68
JO386614.2 - 2 - 3 317 68
JO576367.1 - 2 - 3 702 68

View Full Product Details