Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
CLTRN
Entrez Gene ID
102189658
Gene Name
collectrin, amino acid transport regulator
Gene Aliases
TMEM27
Chromosome Location
Chr.X|NW_017189516: 54621946 - 54671371 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018043778.1 XP_017899267.1 5 - 6 869 80
GenBank mRNA JO409524.2 - 5 - 6 589 80
JO434592.2 - 1 - 2 927 80

View Full Product Details