Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LOC102191731
Entrez Gene ID
102191731
Gene Name
zinc finger and SCAN domain-containing protein 4
Gene Aliases
ZSCAN4
Chromosome Location
Chr.18: 66352648 - 66354897 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005693058.2 XP_005693115.2 2 - 3 553 88

View Full Product Details