Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HAUS5
Entrez Gene ID
102179976
Gene Name
HAUS augmin like complex subunit 5
Gene Aliases
-
Chromosome Location
Chr.18: 47181011 - 47192218 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018063320.1 XP_017918809.1 9 - 10 763 55
GenBank mRNA JT702646.1 - 10 - 11 1185 55

View Full Product Details