Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BTBD1
Entrez Gene ID
102170365
Gene Name
BTB domain containing 1
Gene Aliases
-
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018066349.1 XP_017921838.1 8 - 9 1505 62
XM_018066350.1 XP_017921839.1 7 - 8 1460 62
GenBank mRNA JT762761.1 - 1 - 2 413 62

View Full Product Details