Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CRYM
Entrez Gene ID
100861024
Gene Name
crystallin mu
Gene Aliases
-
Chromosome Location
Chr.25: 18923735 - 18947678 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005697591.3 XP_005697648.1 1 - 2 595 51

View Full Product Details