Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EAPP
Entrez Gene ID
102180085
Gene Name
E2F associated phosphoprotein
Gene Aliases
-
Chromosome Location
Chr.21: 44355581 - 44371258 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018066492.1 XP_017921981.1 4 - 5 555 59

View Full Product Details