Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
GPAT4
Entrez Gene ID
100861270
Gene Name
glycerol-3-phosphate acyltransferase 4
Gene Aliases
AGPAT6
Chromosome Location
Chr.27: 9070923 - 9101550 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018041983.1 XP_017897472.1 5 - 6 1528 66
XM_018041984.1 XP_017897473.1 6 - 7 1569 66
GenBank mRNA JO426877.1 - 5 - 6 1775 66

View Full Product Details