Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ARID3C
Entrez Gene ID
102185273
Gene Name
AT-rich interaction domain 3C
Gene Aliases
-
Chromosome Location
Chr.8: 75814302 - 75821141 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018052839.1 XP_017908328.1 4 - 5 961 93

View Full Product Details