Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DCDC1
Entrez Gene ID
102183892
Gene Name
doublecortin domain containing 1
Gene Aliases
-
Chromosome Location
Chr.15: 20761459 - 20858273 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005709613.3 XP_005709670.2 2 - 3 372 109

View Full Product Details