Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SDC2
Entrez Gene ID
102183082
Gene Name
syndecan 2
Gene Aliases
-
Chromosome Location
Chr.14: 14508815 - 14632178 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA JO438416.2 - 3 - 4 843 109

View Full Product Details