Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Trpgamma
Entrez Gene ID
34991
Gene Name
Transient receptor potential cation channel gamma
Gene Aliases
CG5996, DmTRPgamma, Dmel\CG5996, Dmel_CG5996, TRPgamma, Trp-gamma, dTRPg, dTRPgamma, trp-gamma, trpgamma
Chromosome Location
Chr.2L: 16663454 - 16676701 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0032593
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001144358.3 NP_001137830.2 12 - 13 2632 71
NM_135958.5 NP_609802.1 13 - 14 2675 71
NM_165169.5 NP_723983.1 12 - 13 2632 71

View Full Product Details