Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Nhe1
- Entrez Gene ID
-
33167
- Gene Name
- Na[+]/H[+] hydrogen exchanger 1
- Gene Aliases
-
CG12178, DmNHE1, Dmel\CG12178, Dmel_CG12178, NHE1, nhe1
- Chromosome Location
-
Chr.2L: 103962 - 106732 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0026787
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_134647.4 |
NP_608491.3 |
3 - 4 |
1239 |
64 |
View Full Product Details