Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KdelR
Entrez Gene ID
34427
Gene Name
KDEL receptor
Gene Aliases
BEST:CK00230, CG5183, CK00230, Dmel\CG5183, Dmel_CG5183, ERD2, Erd2, dmErd2, l(2)31Em, l(2)k00311
Chromosome Location
Chr.2L: 10426887 - 10428632 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0267330
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_057948.3 NP_477296.1 2 - 2 1045 145
NM_164927.2 NP_723586.1 1 - 1 1117 145
NM_164928.3 NP_723587.1 2 - 2 1050 145

View Full Product Details