Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ge-1
Entrez Gene ID
34541
Gene Name
CG6181-PA
Gene Aliases
CG6181, Dmel\CG6181, Dmel_CG6181, EDC4, GE-1, Ref(2)Me, dGe-1, ref(2)M
Chromosome Location
Chr.2L: 11095150 - 11100888 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0283682
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_135642.3 NP_609486.2 5 - 6 1374 120
NM_164957.2 NP_723654.1 4 - 5 1457 120
GenBank mRNA BT004479.1 - 5 - 6 1354 120

View Full Product Details