Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- hoe2
- Entrez Gene ID
-
33700
- Gene Name
- hoepel2
- Gene Aliases
-
CG15624, Dmel\CG15624, Dmel_CG15624
- Chromosome Location
-
Chr.2L: 4933047 - 4936450 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0031649
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_135034.3 |
NP_608878.1 |
3 - 4 |
909 |
59 |
View Full Product Details