Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Etl1
- Entrez Gene ID
-
34311
- Gene Name
- CG5899-PA
- Gene Aliases
-
CG5899, Dmel\CG5899, Dmel_CG5899, ETL1, SMARCAD1
- Chromosome Location
-
Chr.2L: 9908830 - 9912608 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0032157
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_135476.4 |
NP_609320.2 |
5 - 6 |
1586 |
70 |
View Full Product Details