Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Prosbeta5R2
Entrez Gene ID
318924
Gene Name
Proteasome beta5 subunit-related 2
Gene Aliases
31742, CG31742, Dmel\CG31742, Dmel_CG31742, anon-AE003657.1, beta5R2
Chromosome Location
Chr.2L: 17990418 - 17991951 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0051742
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001299093.1 NP_001286022.1 1 - 1 867 70
NM_165234.3 NP_724081.1 1 - 1 867 70
GenBank mRNA BT023520.1 - 1 - 1 867 70

View Full Product Details