Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rab32
Entrez Gene ID
35940
Gene Name
CG8024-PB
Gene Aliases
24652026, 24652028, A1Z7S1, CG8024, DRABR1, DmRab32, Dmel\CG8024, Dmel_CG8024, DrabRP1, Ltd, RAB-RP1, RAB7L, Rab-RP1, Rab-RP1/CG8024, Rab-r1, Rab32/RP1, Rab7L1, RabRP1, anon-WO0118547.106, beck, ltd, rab32
Chromosome Location
Chr.2R: 9177864 - 9186342 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0002567
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_165659.2 NP_724764.1 1 - 2 1123 64
NM_165660.2 NP_724765.1 2 - 3 932 64
GenBank mRNA AY069328.1 - 1 - 2 1147 64

View Full Product Details