Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CAP
Entrez Gene ID
36084
Gene Name
CAP-PA
Gene Aliases
CAP/Vinexin, CG18408, CG18409, CG3451, DCAP, Dmel\CG18408, Dmel_CG18408, Rexin, cap, rexin
Chromosome Location
Chr.2R: 10269016 - 10307024 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0033504
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001144164.2 NP_001137636.1 6 - 7 920 65
NM_001144168.2 NP_001137640.1 4 - 5 443 65
NM_001259311.2 NP_001246240.1 5 - 6 491 65
NM_001259312.2 NP_001246241.1 4 - 5 6312 65
NM_001382041.1 NP_001369073.1 6 - 7 1129 65
NM_001382043.1 NP_001369075.1 4 - 5 6642 65
NM_165757.3 NP_724910.1 7 - 8 941 65
NM_165758.2 NP_724911.1 6 - 7 1129 65
GenBank mRNA BT003217.1 - 7 - 8 942 65
BT003435.1 - 6 - 7 901 65

View Full Product Details