Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Flo2
Entrez Gene ID
32425
Gene Name
Flotillin 2
Gene Aliases
CG11547, CG14409, CG32593, DmFlo2, Dmel\CG32593, Dmel_CG32593, FLO2, FLO[Dm]-2, Flo, Flo-2, flo2, mdcds_54791, reggie-1
Chromosome Location
Chr.X: 14839376 - 14933946 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0264078
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001272623.2 NP_001259552.1 2 - 3 194 58
NM_001272624.2 NP_001259553.1 2 - 3 194 58
NM_001272625.2 NP_001259554.1 2 - 3 194 58
NM_001272626.2 NP_001259555.1 2 - 3 535 58
NM_078602.4 NP_511157.2 2 - 3 194 58
NM_167415.3 NP_727797.1 2 - 3 194 58
NM_167416.3 NP_727798.1 2 - 3 306 58
NM_167417.3 NP_727799.1 2 - 3 306 58
GenBank mRNA BT009962.1 - 2 - 3 192 58
BT126133.1 - 2 - 3 173 58

View Full Product Details