Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Fim
Entrez Gene ID
32721
Gene Name
Fimbrin
Gene Aliases
CG8649, Dmel\CG8649, Dmel_CG8649, FIM, fim
Chromosome Location
Chr.X: 17285704 - 17301929 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0024238
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_078661.4 NP_523385.1 4 - 5 2212 64
NM_167564.2 NP_728073.1 5 - 6 2094 64
NM_167565.2 NP_728074.1 4 - 5 2061 64
GenBank mRNA AF053225.1 - 4 - 5 2179 64
AY069342.1 - 4 - 5 2223 64
BT133379.1 - 4 - 5 2035 64
BT133476.1 - 5 - 6 2094 64

View Full Product Details