Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Or24a
Entrez Gene ID
33623
Gene Name
Odorant receptor 24a
Gene Aliases
24D.1, 24a, CG11767, DOR24D.1, DOR48, Dmel\CG11767, Dmel_CG11767, Or24D.1
Chromosome Location
Chr.2L: 4164689 - 4166134 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0026394
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_078746.5 NP_523470.3 4 - 5 1146 99

View Full Product Details