Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Tep3
- Entrez Gene ID
-
34045
- Gene Name
- Thioester-containing protein 3
- Gene Aliases
-
CG7068, Dmel\CG7068, Dmel_CG7068, TEP3, TEP3mel, TEPIII, TepIII, dTEPIII
- Chromosome Location
-
Chr.2L: 7702805 - 7710690 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0041181
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_078783.3 |
NP_523507.2 |
3 - 4 |
1082 |
82 |
GenBank mRNA |
AY069050.1 |
- |
3 - 4 |
1048 |
82 |
View Full Product Details