Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
bwa
Entrez Gene ID
250736
Gene Name
brain washing
Gene Aliases
38B.6, CG13969, Dacer, Dmel\CG13969, Dmel_CG13969, dacer
Chromosome Location
Chr.2L: 20066391 - 20073719 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0045064
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001273687.2 NP_001260616.1 2 - 3 769 62
NM_136176.4 NP_610020.1 2 - 3 769 62
GenBank mRNA AY071232.1 - 2 - 3 802 62

View Full Product Details